Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004318.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004318.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004318.hg.1 |
TC13000395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000464141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000464141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
ENST00000464141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004317.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004317.hg.1 |
ENST00000464141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004317.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004317.hg.1 |
TC13000395.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004316.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004316.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004316.hg.1 |
TC13000395.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300153989.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300153989.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300153989.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300153989.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300153989.hg.1 |
TC1300008025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000333219.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000338450.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000375775.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000464141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000464141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
ENST00000464141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_001267728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008025.hg.1 |
NM_198219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300153986.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300153986.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300153986.hg.1 |
ENST00000375774.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300153986.hg.1 |
NM_005537.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300153986.hg.1 |
TC1300008025.hg.1 |
Clariom D TC |