Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015512.4 | NP_056327.4 | 75 - 76 | 12347 | 96 | |
XM_017006129.1 | XP_016861618.1 | 77 - 78 | 15304 | 96 | ||
XM_017006130.1 | XP_016861619.1 | 76 - 77 | 15234 | 96 | ||
XM_017006131.1 | XP_016861620.1 | 76 - 77 | 15178 | 96 | ||
GenBank mRNA | AB037831.3 | - | 75 - 76 | 12347 | 96 | |
AB290163.1 | - | 74 - 75 | 12542 | 96 | ||
AK023766.1 | - | 15 - 16 | 2357 | 96 | ||
AK074130.2 | - | 22 - 23 | 3872 | 96 | ||
AL117428.1 | - | 7 - 8 | 1066 | 96 | ||
AY221994.1 | - | 31 - 32 | 5390 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |