Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012488.hg.1 |
TC09000933.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012488.hg.1 |
ENST00000380496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012488.hg.1 |
ENST00000380502.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012488.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008805.hg.1 |
ENST00000433254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008805.hg.1 |
ENST00000433254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008805.hg.1 |
ENST00000433254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008805.hg.1 |
ENST00000433254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
ENST00000415524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009645.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166056.hg.1 |
TC0900009645.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC2000008805.hg.1 |
ENST00000433254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008805.hg.1 |
ENST00000433254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008805.hg.1 |
ENST00000433254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008805.hg.1 |
ENST00000433254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
ENST00000380496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
ENST00000380502.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
NM_001270890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
NM_017645.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166054.hg.1 |
TC0900009645.hg.1 |
Clariom D TC |