Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034003.hg.1 |
TC03001887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034003.hg.1 |
ENST00000296059.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034003.hg.1 |
ENST00000470080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034003.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034003.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034004.hg.1 |
TC03001887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034004.hg.1 |
ENST00000296059.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034004.hg.1 |
ENST00000470080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034004.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034004.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034010.hg.1 |
TC03001887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034010.hg.1 |
ENST00000296059.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034010.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000468398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000468398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000468398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034007.hg.1 |
TC03001887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034007.hg.1 |
ENST00000296059.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034007.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000468398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000468398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000468398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012770.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000468398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000468398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195466.hg.1 |
TC0300012770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
ENST00000470080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
NM_001184723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195467.hg.1 |
TC0300012770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
ENST00000474754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
ENST00000491182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195471.hg.1 |
TC0300012770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300195474.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195474.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195474.hg.1 |
ENST00000296059.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195474.hg.1 |
NM_138786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195474.hg.1 |
TC0300012770.hg.1 |
Clariom D TC |