Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005366.1 | NP_001005366.1 | 17 - 18 | 2811 | 67 | |
NM_032590.4 | NP_115979.3 | 18 - 19 | 2715 | 67 | ||
XM_005253955.3 | XP_005254012.1 | 18 - 19 | 2925 | 67 | ||
XM_005253956.3 | XP_005254013.1 | 18 - 19 | 2522 | 67 | ||
XM_005253961.4 | XP_005254018.1 | 7 - 8 | 1010 | 67 | ||
XM_011538867.2 | XP_011537169.1 | 18 - 19 | 3977 | 67 | ||
XM_011538868.2 | XP_011537170.1 | 18 - 19 | 3973 | 67 | ||
XM_011538869.2 | XP_011537171.1 | 18 - 19 | 3975 | 67 | ||
XM_011538875.2 | XP_011537177.1 | 7 - 8 | 1019 | 67 | ||
GenBank mRNA | AB031230.1 | - | 8 - 9 | 1183 | 67 | |
AJ459424.1 | - | 18 - 19 | 2687 | 67 | ||
AK027440.1 | - | 8 - 9 | 1104 | 67 | ||
AK027692.1 | - | 5 - 6 | 620 | 67 | ||
AK127328.1 | - | 17 - 18 | 2811 | 67 | ||
AK299827.1 | - | 7 - 8 | 1029 | 67 | ||
AK316470.1 | - | 7 - 8 | 1022 | 67 | ||
BC008735.2 | - | 4 - 5 | 485 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |