Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000468981.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000468981.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000468981.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017736.hg.1 |
TC09001421.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017736.hg.1 |
ENST00000374853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017736.hg.1 |
ENST00000374855.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017736.hg.1 |
ENST00000430164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017736.hg.1 |
ENST00000468981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017736.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017739.hg.1 |
TC09001421.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017739.hg.1 |
ENST00000374853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017739.hg.1 |
ENST00000374855.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017739.hg.1 |
ENST00000430164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017739.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017738.hg.1 |
TC09001421.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017738.hg.1 |
ENST00000374853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017738.hg.1 |
ENST00000374855.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017738.hg.1 |
ENST00000430164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017738.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017737.hg.1 |
TC09001421.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017737.hg.1 |
ENST00000374853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017737.hg.1 |
ENST00000374855.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017737.hg.1 |
ENST00000430164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017737.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
ENST00000468981.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
ENST00000468981.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175201.hg.1 |
TC0900011033.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900175202.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175202.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175202.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175202.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175202.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175202.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175202.hg.1 |
TC0900011033.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900175204.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175204.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175204.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175204.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175204.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175204.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175204.hg.1 |
TC0900011033.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000468981.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000468981.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000468981.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011033.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175203.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175203.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175203.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175203.hg.1 |
ENST00000374855.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175203.hg.1 |
ENST00000616752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175203.hg.1 |
NM_000035.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175203.hg.1 |
TC0900011033.hg.1 |
Clariom D TC |