Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013084.hg.1 |
TC07000831.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013084.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013084.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000473068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000473068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000473068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
ENST00000473068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700165549.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700165549.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700165549.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700165549.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700165549.hg.1 |
TC0700009222.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000285928.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000473068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000473068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000473068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
ENST00000473068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009222.hg.1 |
NM_144648.2 |
Refseq |