Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000547219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000547414.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000548154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000549092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000549921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000550169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000550316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000550389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000550937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001042555.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278354.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_006654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000547219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000547414.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000548154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000549092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000549921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000550169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000550316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000550389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
ENST00000550937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001042555.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278354.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_001278357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
NM_006654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200159645.hg.1 |
TC1200008142.hg.1 |
Clariom D TC |