Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024190.hg.1 |
ENST00000321424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024190.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024190.hg.1 |
TC19001780.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024189.hg.1 |
ENST00000321424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024189.hg.1 |
ENST00000340550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024189.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024189.hg.1 |
TC19001780.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024191.hg.1 |
ENST00000321424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024191.hg.1 |
ENST00000340550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024191.hg.1 |
ENST00000430817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024191.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024191.hg.1 |
TC19001780.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
TC1900011257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194779.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194780.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194780.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194780.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194780.hg.1 |
TC1900011257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194780.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194780.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194780.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
TC1900011257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194782.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
ENST00000597352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011257.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
NM_014442.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
TC1900011257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
ENST00000321424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
ENST00000340550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
ENST00000430817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194783.hg.1 |
NM_014442.2 |
Refseq |