Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001303029.1 | NP_001289958.1 | 6 - 7 | 1347 | 67 | |
NM_007183.3 | NP_009114.1 | 5 - 6 | 1383 | 67 | ||
GenBank mRNA | AF053719.1 | - | 5 - 6 | 1327 | 67 | |
AK025067.1 | - | 5 - 6 | 1344 | 67 | ||
AK223511.1 | - | 5 - 6 | 1344 | 67 | ||
BC000081.2 | - | 5 - 6 | 1349 | 3504300 | 67 | |
CA947983.1 | - | 3 - 4 | 67 | |||
FN421477.1 | - | 6 - 7 | 1347 | 67 | ||
Z98265.1 | - | 5 - 6 | 1347 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |