Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013466.hg.1 |
TC08002612.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013466.hg.1 |
ENST00000358689.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013466.hg.1 |
ENST00000417069.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013466.hg.1 |
NM_001128930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013466.hg.1 |
NM_004901.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000356206.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000358689.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000417069.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000520936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000523958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
NM_001128930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
NM_004901.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000356206.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000358689.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000417069.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000520936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
ENST00000523958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
NM_001128930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
NM_004901.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167136.hg.1 |
TC0800009878.hg.1 |
Clariom D TC |