Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000460159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000460159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000460159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000460159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025058.hg.1 |
TC06002009.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025058.hg.1 |
ENST00000330550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025058.hg.1 |
ENST00000368919.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025058.hg.1 |
ENST00000439654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025058.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000424139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000434949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000437378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000460159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000460159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000460159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000460159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
ENST00000461487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012833.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
ENST00000330550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
ENST00000368919.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
ENST00000451557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
NM_033125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188617.hg.1 |
TC0600012833.hg.1 |
Clariom D TC |