Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000462169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000462169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000462169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000485549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000485549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000485549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016447.hg.1 |
ENST00000341546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016447.hg.1 |
ENST00000393992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016447.hg.1 |
ENST00000409037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016447.hg.1 |
ENST00000409509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016447.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016447.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016447.hg.1 |
TC16001183.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000447579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000462169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000462169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000462169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000485549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000485549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000485549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010622.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000393992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000409037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000409509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000424285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000433915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
ENST00000629962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
NM_001004055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
NM_012163.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600181762.hg.1 |
TC1600010622.hg.1 |
Clariom D TC |