Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018051.hg.1 |
TC03001018.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018051.hg.1 |
ENST00000265028.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018051.hg.1 |
ENST00000439351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018051.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000464877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000464877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000464877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000464877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206087.hg.1 |
TC0300013911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000265028.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000439351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000464877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000464877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000464877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000464877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
ENST00000495390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013911.hg.1 |
NM_016306.5 |
Refseq |