Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005648.hg.1 |
ENST00000355868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005648.hg.1 |
ENST00000361680.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005648.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005648.hg.1 |
TC19000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157065.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157065.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157065.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157065.hg.1 |
TC1900007764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157065.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157065.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157065.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000355868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000361680.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
ENST00000592765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007764.hg.1 |
NM_173479.3 |
Refseq |