Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018215.hg.1 |
ENST00000225873.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018215.hg.1 |
ENST00000424525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018215.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018215.hg.1 |
TC17001374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018214.hg.1 |
ENST00000225873.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018214.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018214.hg.1 |
TC17001374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000585380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000585380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000585380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000585380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000585380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000585380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000585380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000585380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010436.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700181873.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181873.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181873.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700181873.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181873.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181873.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700181873.hg.1 |
TC1700010436.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
ENST00000225873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
ENST00000586663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
ENST00000613219.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
NM_000286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700181875.hg.1 |
TC1700010436.hg.1 |
Clariom D TC |