Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000389554.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000410035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000411412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000411412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000411412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000411412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000411412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000411412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000411412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000411412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000463544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000477804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000477804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000477804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000477804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000477804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000477804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000489530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000489530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000489530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000505579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
ENST00000505579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016555.hg.1 |
NM_006593.3 |
Refseq |