Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
ENST00000260229.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012125.hg.1 |
NM_022122.2 |
Refseq |