Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009645.hg.1 |
ENST00000339841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009645.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009645.hg.1 |
TC19000696.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009648.hg.1 |
ENST00000339841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009648.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009648.hg.1 |
TC19000696.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009647.hg.1 |
ENST00000339841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009647.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009647.hg.1 |
TC19000696.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009646.hg.1 |
ENST00000339841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009646.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009646.hg.1 |
TC19000696.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009649.hg.1 |
ENST00000339841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009649.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009649.hg.1 |
TC19000696.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900165009.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165009.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165009.hg.1 |
TC1900008456.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165009.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165009.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
TC1900008456.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165015.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165016.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165016.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165016.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165016.hg.1 |
TC1900008456.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165016.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165016.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165016.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
TC1900008456.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165017.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000593782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
ENST00000619003.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008456.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
NM_022142.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
TC1900008456.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
ENST00000339841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
ENST00000596043.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
ENST00000597519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165012.hg.1 |
NM_022142.4 |
Refseq |