Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049520.hg.1 |
TC01003220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049520.hg.1 |
ENST00000368908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049520.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049519.hg.1 |
TC01003220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049519.hg.1 |
ENST00000368908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049519.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049519.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049519.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049521.hg.1 |
TC01003220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049521.hg.1 |
ENST00000368908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049521.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218299.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218299.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218299.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218299.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218299.hg.1 |
TC0100015774.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015774.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218297.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218297.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218297.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218297.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218297.hg.1 |
TC0100015774.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100218298.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218298.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218298.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218298.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218298.hg.1 |
TC0100015774.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
ENST00000368908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
ENST00000440902.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
NM_001136543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
NM_212551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218296.hg.1 |
TC0100015774.hg.1 |
Clariom D TC |