Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144668.5 | NP_653269.3 | 19 - 20 | 3161 | 102 | |
GenBank mRNA | AL833930.1 | - | 17 - 18 | 2449 | 102 | |
BC028421.2 | - | 19 - 20 | 3159 | 4826893 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |