Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144668.5 | NP_653269.3 | 20 - 21 | 3398 | 72 | |
GenBank mRNA | AL833930.1 | - | 18 - 19 | 2686 | 72 | |
BC028421.2 | - | 20 - 21 | 3396 | 4826893 | 72 | |
CA503090.1 | - | 2 - 3 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |