Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006475.2 | NP_006466.2 | 16 - 17 | 2122 | 133 | |
XM_005266231.2 | XP_005266288.1 | 16 - 17 | 2117 | 133 | ||
XM_017020355.1 | XP_016875844.1 | 16 - 17 | 2117 | 133 | ||
XM_017020356.1 | XP_016875845.1 | 16 - 17 | 2117 | 133 | ||
GenBank mRNA | D13666.1 | - | 16 - 17 | 2015 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |