Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
TC01001661.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
ENST00000295624.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
ENST00000367295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
ENST00000367296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
ENST00000367297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
ENST00000367300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
ENST00000367301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
ENST00000367302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
ENST00000391966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
NM_001167738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025735.hg.1 |
NM_020443.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
TC01001661.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
ENST00000295624.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
ENST00000367295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
ENST00000367296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
ENST00000367297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
ENST00000367300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
ENST00000367302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
ENST00000391966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
NM_001167738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025736.hg.1 |
NM_020443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
ENST00000367295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
ENST00000367296.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
ENST00000367302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
NM_001167738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
NM_020443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
ENST00000367295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
ENST00000367296.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
ENST00000367302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
NM_001167738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
NM_020443.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240266.hg.1 |
TC0100018338.hg.1 |
Clariom D TC |