Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF548116.1 | - | 14 - 15 | 1827 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons |