Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014365.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014365.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014365.hg.1 |
TC11001139.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014364.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014364.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014364.hg.1 |
TC11001139.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100170320.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170320.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170320.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170320.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170320.hg.1 |
TC1100009394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009394.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170319.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170319.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170319.hg.1 |
ENST00000284288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170319.hg.1 |
NM_052959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170319.hg.1 |
TC1100009394.hg.1 |
Clariom D TC |