Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000542305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000542305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000542305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000542305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026633.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026633.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026633.hg.1 |
TC11002229.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000542305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000542305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000542305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000542305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
ENST00000544938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012124.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193164.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193164.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193164.hg.1 |
ENST00000260228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193164.hg.1 |
NM_004771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193164.hg.1 |
TC1100012124.hg.1 |
Clariom D TC |