Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011199.hg.1 |
TC04000808.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011199.hg.1 |
ENST00000264433.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011199.hg.1 |
ENST00000379346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011199.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000505130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000505130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000505445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000505445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000505445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000505445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162902.hg.1 |
TC0400009137.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000264433.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000504715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000505130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000505130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000505445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000505445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000505445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000505445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
ENST00000512986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009137.hg.1 |
NM_020840.2 |
Refseq |