Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC85A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080433.1 | NP_001073902.1 | 3 - 4 | 1820 | 130 | |
GenBank mRNA | AB067499.2 | - | 3 - 4 | 1904 | 130 | |
BC131557.1 | - | 3 - 4 | 492 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |