Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032656.3 | NP_116045.2 | 27 | 4346 | 125 | |
GenBank mRNA | AB040950.1 | - | 23 | 3914 | 125 | |
AK123968.1 | - | 6 | 2554 | 125 | ||
BC004463.1 | - | 1 | 841 | 125 | ||
BC009913.2 | - | 1 | 841 | 125 | ||
BC037964.1 | - | 27 | 5736582 | 125 | ||
BC071824.1 | - | 2 | 937 | 125 | ||
BU633279.1 | - | 1 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |