Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Hs01590968_m1
See other MBNL1 GE Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01590968_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: MBNL1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001314057.1
NP_001300986.1
9 - 10
1684
179
NM_021038.4
NP_066368.2
7 - 8
2415
179
NM_207295.1
NP_997178.1
5 - 6
1586
179
XM_005247457.4
XP_005247514.1
9 - 10
2037
179
XM_005247458.4
XP_005247515.1
9 - 10
3291
179
XM_005247460.4
XP_005247517.1
9 - 10
2000
179
XM_005247462.4
XP_005247519.1
8 - 9
1983
179
XM_005247464.4
XP_005247521.1
8 - 9
2095
179
XM_005247474.3
XP_005247531.1
7 - 8
955
179
XM_005247475.3
XP_005247532.1
8 - 9
1278
179
XM_005247476.4
XP_005247533.1
6 - 7
991
179
XM_005247477.4
XP_005247534.1
7 - 8
1044
179
XM_006713639.1
XP_006713702.1
9 - 10
2019
179
XM_011512843.2
XP_011511145.1
8 - 9
1833
179
XM_011512845.2
XP_011511147.1
7 - 8
1779
179
XM_011512847.2
XP_011511149.1
6 - 7
1742
179
XM_011512848.2
XP_011511150.1
9 - 10
1641
179
XM_017006423.1
XP_016861912.1
8 - 9
2288
179
XM_017006424.1
XP_016861913.1
8 - 9
2115
179
XM_017006427.1
XP_016861916.1
7 - 8
2208
179
XM_017006428.1
XP_016861917.1
7 - 8
1872
179
XM_017006429.1
XP_016861918.1
7 - 8
1955
179
XM_017006435.1
XP_016861924.1
8 - 9
1085
179
XM_017006439.1
XP_016861928.1
7 - 8
1224
179
XM_017006440.1
XP_016861929.1
6 - 7
958
179
GenBank mRNA
AF395876.1
-
5 - 6
797
179
AF401998.1
-
6 - 7
1001
179
AJ308400.1
-
6 - 7
1030
179
AK304089.1
-
6 - 7
764
179
BC043493.1
-
7 - 8
1920
6044671
179
BC050535.1
-
9 - 10
1205
6042311
179
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene MBNL1
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0