Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011541209.2 | XP_011539511.1 | 7 - 8 | 861 | 153 | |
XM_011541210.2 | XP_011539512.1 | 7 - 8 | 861 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Homo sapiens Inventoried Probe spans exons |