Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000348899.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000348899.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000348899.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000348899.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000403974.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
ENST00000431453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013336.hg.1 |
NR_002217.1 |
Refseq |