Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011632.hg.1 |
ENST00000340363.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011632.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011632.hg.1 |
TC17000895.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011633.hg.1 |
ENST00000340363.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011633.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011633.hg.1 |
TC17000895.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168230.hg.1 |
TC1700009037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700168228.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168228.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168228.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168228.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168228.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168228.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168228.hg.1 |
TC1700009037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700168226.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168226.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168226.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168226.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168226.hg.1 |
TC1700009037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000340363.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000451352.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000586999.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
ENST00000591995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009037.hg.1 |
NM_001163075.1 |
Refseq |