Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029754.hg.1 |
TC01001914.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029754.hg.1 |
ENST00000366652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029754.hg.1 |
ENST00000366653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029754.hg.1 |
ENST00000444294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029754.hg.1 |
ENST00000449018.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029754.hg.1 |
NM_001004342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
ENST00000366653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
ENST00000444294.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
ENST00000449018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
NM_001004342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
NM_001300889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
ENST00000366653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
ENST00000444294.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
ENST00000449018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
NM_001004342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
NM_001300889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188205.hg.1 |
TC0100011957.hg.1 |
Clariom D TC |