Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024452.2 | NP_001019623.2 | 2 - 3 | 623 | 169 | |
NM_001172285.1 | NP_001165756.1 | 2 - 3 | 623 | 169 | ||
GenBank mRNA | AK309010.1 | - | 2 - 3 | 623 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Detect Genomic DNA Homo sapiens Probe spans exons |