Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022389.hg.1 |
TC09002927.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022389.hg.1 |
ENST00000341206.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022389.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000471509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000480347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000494091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
ENST00000497749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012289.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186920.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186920.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186920.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186920.hg.1 |
ENST00000341206.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186920.hg.1 |
ENST00000476567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186920.hg.1 |
NM_198946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186920.hg.1 |
TC0900012289.hg.1 |
Clariom D TC |