Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000595583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000598543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000611674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000617846.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000620710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623438.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
ENST00000623806.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_001282505.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011135.hg.1 |
NM_175743.2 |
Refseq |