Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021832.hg.1 |
ENST00000342234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021832.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021832.hg.1 |
TC17001629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021831.hg.1 |
ENST00000342234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021831.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021831.hg.1 |
TC17001629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021827.hg.1 |
ENST00000342234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021827.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021827.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021827.hg.1 |
TC17001629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021826.hg.1 |
ENST00000342234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021826.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021826.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021826.hg.1 |
TC17001629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021829.hg.1 |
ENST00000342234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021829.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021829.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021829.hg.1 |
TC17001629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021828.hg.1 |
ENST00000342234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021828.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021828.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021828.hg.1 |
TC17001629.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700188191.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188191.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188191.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188191.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188191.hg.1 |
TC1700010962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188187.hg.1 |
TC1700010962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700188190.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188190.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188190.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188190.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188190.hg.1 |
TC1700010962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188186.hg.1 |
TC1700010962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010962.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188185.hg.1 |
TC1700010962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
ENST00000342234.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
ENST00000536300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
NM_001258248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
NM_199262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188184.hg.1 |
TC1700010962.hg.1 |
Clariom D TC |