Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015228.hg.1 |
TC06001272.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015228.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015228.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015228.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015229.hg.1 |
TC06001272.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015229.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015229.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015229.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015231.hg.1 |
TC06001272.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015231.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015231.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015233.hg.1 |
TC06001272.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015233.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173505.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173505.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173505.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173505.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173505.hg.1 |
TC0600010807.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600173507.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173507.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173507.hg.1 |
TC0600010807.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600173502.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173502.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173502.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173502.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173502.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173502.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173502.hg.1 |
TC0600010807.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010807.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173503.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173503.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173503.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173503.hg.1 |
ENST00000472091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173503.hg.1 |
ENST00000542862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173503.hg.1 |
NM_207582.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173503.hg.1 |
TC0600010807.hg.1 |
Clariom D TC |