Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013490.hg.1 |
ENST00000329267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013490.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013490.hg.1 |
TC19000989.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
TC1900009104.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172310.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
TC1900009104.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172311.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000329267.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
ENST00000613411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_001308209.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009104.hg.1 |
NM_214710.4 |
Refseq |