Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005255956.4 | XP_005256013.1 | 1 - 2 | 188 | 146 | |
GenBank mRNA | AF495759.1 | - | 1 - 2 | 74 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Inventoried Probe spans exons |