Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111909.1 | 2 - 3 | 126 | 88 | ||
GenBank mRNA | BU154601.1 | - | 2 - 3 | 109 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons |