Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009351.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009351.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009351.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009351.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009351.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009351.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009351.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009351.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900164295.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900164295.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900164295.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900164295.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900164295.hg.1 |
TC0900009351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009351.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009351.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009351.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009351.hg.1 |
ENST00000382387.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009351.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009351.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009351.hg.1 |
NM_152569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009351.hg.1 |
NM_152569.2 |
Refseq |