Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010917.hg.1 |
TC06000932.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010917.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010917.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009573.hg.1 |
ENST00000446295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010916.hg.1 |
TC06000932.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010916.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010916.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006609.hg.1 |
ENST00000423502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006609.hg.1 |
ENST00000423502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006609.hg.1 |
ENST00000423502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006609.hg.1 |
ENST00000423502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010915.hg.1 |
TC06000932.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010915.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010915.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010918.hg.1 |
TC06000932.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010918.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010918.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010909.hg.1 |
TC06000932.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010909.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009575.hg.1 |
ENST00000401550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006573.hg.1 |
ENST00000417358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006573.hg.1 |
ENST00000575845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006573.hg.1 |
ENST00000575845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010914.hg.1 |
TC06000932.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010914.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010914.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000430708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009575.hg.1 |
ENST00000401550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163617.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163617.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163617.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163617.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163617.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600163619.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163619.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163619.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163619.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163619.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600163615.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163615.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163615.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163615.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163615.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600163609.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163609.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163609.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009573.hg.1 |
ENST00000446295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163621.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163621.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163621.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163621.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163621.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600163620.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163620.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163620.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163620.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163620.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600163622.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163622.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163622.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163622.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163622.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006609.hg.1 |
ENST00000423502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006609.hg.1 |
ENST00000423502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006609.hg.1 |
ENST00000423502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006609.hg.1 |
ENST00000423502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006573.hg.1 |
ENST00000417358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006573.hg.1 |
ENST00000575845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006573.hg.1 |
ENST00000575845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163614.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163614.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163614.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163614.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163614.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000430708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |