Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005255859.4 | XP_005255916.3 | 1 | 1055 | 83 | |
XM_005255860.3 | XP_005255917.3 | 1 | 1062 | 83 | ||
XM_005255861.4 | XP_005255918.3 | 1 | 1055 | 83 | ||
XM_017023084.1 | XP_016878573.1 | 1 | 1227 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |