Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541251 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600013 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541277 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541244 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600001 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541269 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541229 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541235 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600003 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600014 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541225 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600038 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541246 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600017 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20599998 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541262 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541220 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541287 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541261 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541271 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541265 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541228 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541276 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600019 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600020 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541247 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20599999 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541281 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541268 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541286 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541242 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20599991 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600008 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600000 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541274 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541219 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541273 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541275 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600042 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600010 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541221 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541288 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600015 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541234 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541263 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541266 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00541270 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600043 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20600021 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
133558 |
AK124749.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
133558 |
AK125159.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
133558 |
AL137278.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
133558 |
AL157492.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
133558 |
NM_020431.1 |
refseq_mrna_human_NM |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P126031 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P126031 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
37294 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
11146 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P10344168 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P126031 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P126031 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600042 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541273 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600013 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541281 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541270 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541261 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600019 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600043 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541271 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541277 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600045 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600038 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600017 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541269 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541276 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541286 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541266 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600020 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541274 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541268 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600015 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541263 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541247 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541286 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600015 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600013 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600042 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541261 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600043 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600017 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541268 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600038 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541277 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541276 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541263 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541270 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541271 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541247 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600045 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541273 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541274 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541281 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600020 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541266 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00541269 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20600019 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P126031 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541253 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541286 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541263 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541256 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541279 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541268 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541248 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541250 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541258 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541247 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541281 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541261 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541275 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541273 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541270 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541266 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00541284 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P126031 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541270 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541273 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541236 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541263 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541279 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541268 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541238 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541246 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541243 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541230 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541221 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541240 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541248 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541286 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541224 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541227 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541258 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541256 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541281 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541284 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541266 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541232 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541253 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541275 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541239 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541250 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541235 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00541288 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P35326638 |
- |
- |