Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004951.hg.1 |
TC08000334.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004951.hg.1 |
ENST00000522175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004951.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004951.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007466.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
ENST00000519951.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
ENST00000522175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
NM_001002920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
NM_001005365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151793.hg.1 |
TC0800007466.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010341.hg.1 |
ENST00000509729.3 |
Ensembl |