Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009583.hg.1 |
TC02000607.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009583.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009583.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160126.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160126.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160126.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160126.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160126.hg.1 |
TC0200008614.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
ENST00000376865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008614.hg.1 |
NM_001011717.1 |
Refseq |