Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146421.2 | NP_666533.1 | 5 - 6 | 437 | 163 | |
GenBank mRNA | AY532927.1 | - | 5 - 6 | 359 | 163 | |
BC024005.2 | - | 5 - 6 | 393 | 4823313 | 163 | |
KJ534858.1 | - | 5 - 6 | 359 | 163 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |